Assay Details
Target Gene Details
Entrez Gene ID: | 56647 |
Gene Name: | BRCA2 and CDKN1A interacting protein |
Gene Aliases: |
TOK-1, TOK1 |
Location: |
Chr.10:125823535-125853695 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCCIP | NM_016567.3 | NP_057651.1 | ||
| NM_078469.2 | NP_510869.1 | |||
| AB040450.1 | BAA92927.1 | |||
| AK055691.1 | ||||
| AK300767.1 | ||||
| AK304529.1 | ||||
| AL834458.1 | CAD39118.1 | |||
| AY064248.1 | AAL55439.1 | |||
| BM974496.1 |
Target Gene Details
Entrez Gene ID: | 55760 |
Gene Name: | DEAH-box helicase 32 (putative) |
Gene Aliases: |
DDX32, DHLP1 |
Location: |
Chr.10:125836337-125896483 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX32 | NM_018180.2 | 11 | 2777 | NP_060650.2 |
| XM_017016404.1 | 13 | 3109 | XP_016871893.1 | |
| XM_017016405.1 | 12 | 2799 | XP_016871894.1 | |
| AF427340.1 | 11 | 2778 | AAL26550.1 | |
| AK001556.1 | 11 | 2571 | BAA91754.1 | |
| AK001751.1 | 11 | 2750 | BAA91882.1 | |
| AK024869.1 | 9 | 1593 | BAB15029.1 | |
| AL162051.1 | 9 | 1575 | CAB82394.1 | |
| AY064250.1 | 11 | 2745 | AAL55441.1 | |
| BC002473.2 | 11 | 2313 | ||
| BC037925.1 | 8 | 2096 | AAH37925.1 | |
| BC068471.1 | 12 | 2774 | AAH68471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8737 | Chr.10:125833274 - 125836965 on Build GRCh38 | Loss |
|
| nsv428241 | Chr.10:125765331 - 125928101 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map