Assay Details
Target Gene Details
Entrez Gene ID: | 81577 |
Gene Name: | glucose-fructose oxidoreductase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.16:67674533-67719370 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFOD2 | NM_001243650.1 | 2 | 430 | NP_001230579.1 |
| NM_030819.3 | 2 | 430 | NP_110446.3 | |
| NR_027398.1 | ||||
| XM_017023738.1 | 2 | 380 | XP_016879227.1 | |
| AK074382.1 | 3 | 383 | BAB85066.1 | |
| AK225997.1 | 2 | 400 | ||
| AK307280.1 | 2 | 384 | ||
| AY358169.1 | 1 | 140 | AAQ88536.1 | |
| BC000757.2 | AAH00757.1 | |||
| BC002943.1 | AAH02943.2 | |||
| BC107585.1 | 2 | 380 | AAI07586.1 | |
| BI826603.1 | 2 | 481 | ||
| DC368734.1 | 2 | 430 | ||
| DC381447.1 | 2 | 385 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map