Assay Details
Target Gene Details
Entrez Gene ID: | 8314 |
Gene Name: | BRCA1 associated protein 1 |
Gene Aliases: |
HUCEP-13, UCHL2, hucep-6 |
Location: |
Chr.3:52401004-52410105 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAP1 | NM_004656.3 | 17 | 2558 | NP_004647.1 |
| XM_011534149.2 | 17 | 2623 | XP_011532451.1 | |
| XM_011534150.2 | 17 | 2578 | XP_011532452.1 | |
| XM_011534151.2 | 17 | 2569 | XP_011532453.1 | |
| XM_011534152.1 | 17 | 2509 | XP_011532454.1 | |
| XM_017007303.1 | 17 | 2500 | XP_016862792.1 | |
| AB002534.1 | BAB46926.1 | |||
| AF045581.1 | 17 | 2370 | AAC15970.1 | |
| AK092725.1 | 5 | 693 | ||
| AK094499.1 | 6 | 1709 | ||
| AK225494.1 | 9 | 1529 | ||
| AK292608.1 | ||||
| AK295395.1 | 5 | 903 | ||
| AK307274.1 | 5 | 806 | ||
| AY130008.1 | 11 | 1843 | AAN05092.1 | |
| BC001596.1 | 17 | 2446 | AAH01596.1 | |
| BX647318.1 | 5 | 530 | ||
| D87462.2 | 17 | 2455 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834696 | Chr.3:52283462 - 52445521 on Build GRCh38 | Loss |
|
| nsv834697 | Chr.3:52366562 - 52545401 on Build GRCh38 | Loss |
|
| nsv954859 | Chr.3:52277285 - 52410684 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map