Assay Details
Target Gene Details
Entrez Gene ID: | 79758 |
Gene Name: | dehydrogenase/reductase 12 |
Gene Aliases: |
SDR40C1 |
Location: |
Chr.13:51754856-51804206 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS12 | NM_001270424.1 | NP_001257353.1 | ||
| NM_024705.2 | NP_078981.1 | |||
| XM_005266527.3 | XP_005266584.1 | |||
| XM_017020750.1 | XP_016876239.1 | |||
| XM_017020751.1 | XP_016876240.1 | |||
| XM_017020754.1 | XP_016876243.1 | |||
| AA831151.1 | ||||
| AK023701.1 | BAB14646.1 | |||
| BC026024.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
| nsv832612 | Chr.13:51602847 - 51788389 on Build GRCh38 | Gain |
|
| dgv3146n54 | Chr.13:51728494 - 51831115 on Build GRCh38 | Loss |
|
| dgv1652n100 | Chr.13:51747428 - 52069788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map