Assay Details
Target Gene Details
Entrez Gene ID: | 55004 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 1 |
Gene Aliases: |
C11orf59, PDRO, Ragulator1, p18, p27RF-Rho |
Location: |
Chr.11:72097292-72103387 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR1 | NM_017907.2 | NP_060377.1 | ||
| AF289583.1 | AAL55767.1 | |||
| AK000632.1 | BAA91297.1 | |||
| AK300681.1 | ||||
| BC001706.1 | AAH01706.1 | |||
| BX381569.2 | ||||
| CR457247.1 | CAG33528.1 |
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTOMT | NM_001145308.4 | NP_001138780.1 | ||
| NM_001145309.3 | NP_001138781.1 | |||
| NM_001145310.3 | NP_001138782.1 | |||
| AF289588.1 | AAL55772.1 | |||
| AK302772.1 | ||||
| EU627069.1 | ||||
| EU627070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv384 | Chr.11:72074236 - 72106174 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map