Assay Details
Target Gene Details
Entrez Gene ID: | 292 |
Gene Name: | solute carrier family 25 member 5 |
Gene Aliases: |
2F1, AAC2, ANT2, T2, T3 |
Location: |
Chr.X:119468400-119471396 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A5 | NM_001152.4 | NP_001143.2 | ||
| AK315292.1 | ||||
| BC056160.1 | AAH56160.1 | |||
| BC068199.1 | AAH68199.1 | |||
| DR759917.1 | ||||
| J02683.1 | AAA35579.1 |
Target Gene Details
Entrez Gene ID: | 100303728 |
Gene Name: | SLC25A5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:119466033-119469120 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A5-AS1 | NR_028443.1 | 1 | 468 | |
| NR_134914.1 | ||||
| AI380428.1 | 1 | 465 | ||
| AI471629.1 | 1 | 468 | ||
| AK092094.1 | 1 | 444 | BAC03803.1 |
More Information
Set Membership: |
|

Genomic Map