Assay Details
Target Gene Details
Entrez Gene ID: | 105370489 |
Gene Name: | uncharacterized LOC105370489 |
Gene Aliases: |
- |
Location: |
Chr.14:50821797-50823509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105370489 | NR_135258.1 | |||
| AI243123.1 | ||||
| HY010266.1 |
Target Gene Details
Entrez Gene ID: | 51199 |
Gene Name: | ninein |
Gene Aliases: |
SCKL7 |
Location: |
Chr.14:50719763-50831121 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIN | NM_016350.4 | 3 | 215 | NP_057434.4 |
| NM_020921.3 | 3 | 215 | NP_065972.3 | |
| NM_182944.2 | 3 | 215 | NP_891989.2 | |
| NM_182946.1 | 3 | 215 | NP_891991.1 | |
| XM_011536817.1 | 2 | 99 | XP_011535119.1 | |
| XM_011536819.2 | 2 | 99 | XP_011535121.1 | |
| XM_011536822.1 | 2 | 99 | XP_011535124.1 | |
| XM_011536823.2 | XP_011535125.1 | |||
| AB046785.2 | 3 | 140 | BAB13391.2 | |
| AF212162.2 | AAF23015.2 | |||
| AF223937.1 | 3 | 225 | AAK00628.1 | |
| AF223938.1 | 3 | 225 | AAK00629.1 | |
| AF223939.1 | 3 | 225 | AAK00630.1 | |
| AF302773.2 | 3 | 215 | AAG33512.2 | |
| AY736182.1 | 3 | 215 | AAU86898.1 | |
| BC065521.1 | AAH65521.1 | |||
| BC090932.1 | 1 | 59 | AAH90932.1 | |
| CR993064.1 | 3 | 234 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832794 | Chr.14:50739915 - 50959891 on Build GRCh38 | Gain |
|
| nsv1040775 | Chr.14:50676760 - 50853094 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map