Assay Details
Target Gene Details
Entrez Gene ID: | 85007 |
Gene Name: | 5-phosphohydroxy-L-lysine phospho-lyase |
Gene Aliases: |
AGXT2L2, PHLU |
Location: |
Chr.5:178208474-178232822 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHYKPL | NM_001278346.1 | NP_001265275.1 | ||
| NM_153373.3 | NP_699204.1 | |||
| NR_103508.1 | ||||
| NR_103509.1 | ||||
| NR_103510.1 | ||||
| XM_006714925.2 | XP_006714988.1 | |||
| XM_011534669.2 | XP_011532971.1 | |||
| XM_011534670.2 | XP_011532972.1 | |||
| XM_011534671.2 | XP_011532973.1 | |||
| XM_011534672.2 | XP_011532974.1 | |||
| XM_011534674.2 | XP_011532976.1 | |||
| XM_011534676.2 | XP_011532978.1 | |||
| XM_017009996.1 | XP_016865485.1 | |||
| XM_017009997.1 | XP_016865486.1 | |||
| XM_017009998.1 | XP_016865487.1 | |||
| XM_017009999.1 | XP_016865488.1 | |||
| AF318375.1 | AAL55882.1 | |||
| AK023470.1 | ||||
| AK093737.1 | 7 | 1320 | ||
| AK292061.1 | ||||
| BC008009.1 | AAH08009.1 | |||
| BC037567.1 | AAH37567.1 | |||
| BC110335.1 | AAI10336.1 | |||
| HY267487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019404 | Chr.5:178080079 - 178432088 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map