Assay Details
Target Gene Details
Entrez Gene ID: | 90835 |
Gene Name: | coiled-coil domain containing 189 |
Gene Aliases: |
C16orf93 |
Location: |
Chr.16:30757226-30762277 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC189 | NM_001014979.2 | 9 | 1424 | NP_001014979.2 |
| XM_011545976.1 | 8 | 1367 | XP_011544278.1 | |
| XM_011545978.1 | 6 | 1212 | XP_011544280.1 | |
| XM_017023852.1 | 8 | 1652 | XP_016879341.1 | |
| XM_017023853.1 | 7 | 1589 | XP_016879342.1 | |
| XM_017023854.1 | 7 | 1562 | XP_016879343.1 | |
| XM_017023855.1 | 9 | 1556 | XP_016879344.1 | |
| XM_017023856.1 | 8 | 1173 | XP_016879345.1 | |
| XM_017023857.1 | 7 | 1643 | XP_016879346.1 | |
| XM_017023858.1 | 6 | 1492 | XP_016879347.1 | |
| XM_017023859.1 | 7 | 1368 | XP_016879348.1 | |
| XM_017023860.1 | 5 | 827 | XP_016879349.1 | |
| XM_017023861.1 | 8 | 1438 | XP_016879350.1 | |
| XM_017023862.1 | 7 | 1970 | XP_016879351.1 | |
| AL122109.2 | 6 | 2006 | ||
| BC092491.1 | 8 | 1716 | AAH92491.1 | |
| BC128128.1 | 8 | 1080 | ||
| BC128129.1 | 9 | 928 |
Target Gene Details
Entrez Gene ID: | 5261 |
Gene Name: | phosphorylase kinase catalytic subunit gamma 2 |
Gene Aliases: |
GSD9C |
Location: |
Chr.16:30748299-30761176 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHKG2 | NM_000294.2 | 10 | 1878 | NP_000285.1 |
| NM_001172432.1 | NP_001165903.1 | |||
| AB209055.1 | 5 | 1267 | BAD92292.1 | |
| AK293551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469724 | Chr.16:30621124 - 30828942 on Build GRCh38 | Loss |
|
| nsv833188 | Chr.16:30621125 - 30828942 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map