Assay Details
Target Gene Details
Entrez Gene ID: | 10747 |
Gene Name: | mannan binding lectin serine peptidase 2 |
Gene Aliases: |
MAP19, MASP-2, MASP1P1, sMAP |
Location: |
Chr.1:11026523-11047239 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MASP2 | NM_006610.3 | 3 | 354 | NP_006601.2 |
| NM_139208.2 | 3 | 354 | NP_631947.1 | |
| XM_017000097.1 | 3 | 354 | XP_016855586.1 | |
| AB008047.1 | 3 | 348 | BAA78616.1 | |
| AK290823.1 | 3 | 354 | ||
| DA638135.1 | 3 | 354 | ||
| X98400.1 | 3 | 358 | CAA67050.1 | |
| Y09926.1 | 3 | 338 | CAA71059.1 | |
| Y18281.1 | 3 | 343 | CAB50728.1 | |
| Y18282.1 | 2 | 292 | CAB50729.1 | |
| Y18283.1 | 3 | 337 | CAB50730.1 | |
| Y18284.1 | 3 | 343 | CAB50731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832425 | Chr.1:10843788 - 11062918 on Build GRCh38 | Loss |
|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
| nsv953456 | Chr.1:11038744 - 11060143 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map