Assay Details
Target Gene Details
Entrez Gene ID: | 89944 |
Gene Name: | galactosidase beta 1 like 2 |
Gene Aliases: |
MST114, MSTP114 |
Location: |
Chr.11:134331874-134376324 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1L2 | NM_138342.3 | NP_612351.2 | ||
| XM_006718937.1 | XP_006719000.1 | |||
| AF173889.1 | AAQ13636.1 | |||
| AK074703.1 | BAC11149.1 | |||
| AK074855.1 | BAC11247.1 | |||
| AK074947.1 | BAC11309.1 | |||
| AY358497.1 | AAQ88861.1 | |||
| BC008326.1 | AAH08326.1 | |||
| BC040641.1 | AAH40641.1 | |||
| BX640934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4e196 | Chr.11:134304439 - 134744665 on Build GRCh38 | Duplication |
|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| nsv556608 | Chr.11:134348406 - 134876236 on Build GRCh38 | Gain |
|
| dgv1305n100 | Chr.11:134363845 - 134416469 on Build GRCh38 | Gain |
|
| nsv1053071 | Chr.11:134355691 - 134379382 on Build GRCh38 | Loss |
|
| dgv1304n100 | Chr.11:134322594 - 134754120 on Build GRCh38 | Gain |
|
| esv3628169 | Chr.11:134353064 - 134485281 on Build GRCh38 | Gain |
|
| nsv561 | Chr.11:134353547 - 134380498 on Build GRCh38 | Insertion |
|
| esv2760555 | Chr.11:134282136 - 134860776 on Build GRCh38 | Gain+Loss |
|
| nsv521623 | Chr.11:134307381 - 134756993 on Build GRCh38 | Gain |
|
| esv3628170 | Chr.11:134357170 - 134435562 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114723326] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map