Assay Details
Target Gene Details
Entrez Gene ID: | 56132 |
Gene Name: | protocadherin beta 3 |
Gene Aliases: |
PCDH-BETA3 |
Location: |
Chr.5:141100242-141103827 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHB3 | NM_018937.4 | 1 | 2945 | NP_061760.1 |
| AF217755.1 | 1 | 2537 | AAK51622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv428127 | Chr.5:141016768 - 141319173 on Build GRCh38 | Loss |
|
| nsv599906 | Chr.5:141097321 - 141103287 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map