Assay Details
Target Gene Details
Entrez Gene ID: | 4054 |
Gene Name: | latent transforming growth factor beta binding protein 3 |
Gene Aliases: |
DASS, LTBP-3, LTBP2, STHAG6, pp6425 |
Location: |
Chr.11:65538559-65558388 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTBP3 | NM_001130144.2 | 8 | 1680 | NP_001123616.1 |
| NM_001164266.1 | 8 | 1676 | NP_001157738.1 | |
| NM_021070.4 | 8 | 1680 | NP_066548.2 | |
| XM_011545032.2 | 8 | 1862 | XP_011543334.1 | |
| XM_011545033.2 | 8 | 1861 | XP_011543335.1 | |
| XM_017017737.1 | 8 | 1867 | XP_016873226.1 | |
| AF011407.1 | 2 | 232 | AAB64201.1 | |
| AF135960.3 | 8 | 1411 | AAF62352.3 | |
| AF318354.1 | 6 | 712 | AAL55861.1 | |
| AK024477.1 | 8 | 1677 | ||
| AK298858.1 | 3 | 362 | ||
| AK299351.1 | 9 | 1342 | ||
| AK309230.1 | 7 | 882 | ||
| BC136277.1 | 8 | 1486 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map