Assay Details
Target Gene Details
Entrez Gene ID: | 8464 |
Gene Name: | SPT3 homolog, SAGA and STAGA complex component |
Gene Aliases: |
SPT3, SPT3L |
Location: |
Chr.6:44808844-45378051 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT3H | NM_001261823.1 | 10 | 1339 | NP_001248752.1 |
| NM_003599.3 | 11 | 1430 | NP_003590.1 | |
| NM_181356.2 | 13 | 1615 | NP_852001.1 | |
| XM_011514949.2 | 14 | 1522 | XP_011513251.1 | |
| XM_011514950.1 | 12 | 1501 | XP_011513252.1 | |
| XM_011514951.2 | 12 | 5562 | XP_011513253.1 | |
| XM_011514952.2 | 12 | 2548 | XP_011513254.1 | |
| XM_011514953.2 | 12 | 1331 | XP_011513255.1 | |
| XM_011514954.2 | 12 | 1362 | XP_011513256.1 | |
| XM_017011369.1 | 14 | 5750 | XP_016866858.1 | |
| XM_017011370.1 | 12 | 1315 | XP_016866859.1 | |
| XM_017011371.1 | 12 | 4605 | XP_016866860.1 | |
| XM_017011372.1 | 11 | 1319 | XP_016866861.1 | |
| XM_017011373.1 | 12 | 1372 | XP_016866862.1 | |
| XM_017011374.1 | 11 | 1197 | XP_016866863.1 | |
| AK303838.1 | 10 | 1152 | ||
| BC035816.1 | 11 | 1307 | AAH35816.1 | |
| BC050384.1 | 13 | 1497 | AAH50384.1 |
More Information
Set Membership: |
|

Genomic Map