Assay Details
Target Gene Details
Entrez Gene ID: | 4638 |
Gene Name: | myosin light chain kinase |
Gene Aliases: |
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK |
Location: |
Chr.3:123612296-123884302 on Build GRCh38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK | NM_001321309.1 | 33 | 6122 | NP_001308238.1 |
| NM_053025.3 | 34 | 6330 | NP_444253.3 | |
| NM_053026.3 | 33 | 6123 | NP_444254.3 | |
| NM_053027.3 | 33 | 6177 | NP_444255.3 | |
| NM_053028.3 | 32 | 5970 | NP_444256.3 | |
| XM_017006469.1 | 17 | 3606 | XP_016861958.1 | |
| XM_017006470.1 | 16 | 2594 | XP_016861959.1 | |
| XM_017006471.1 | 16 | 2647 | XP_016861960.1 | |
| XM_017006472.1 | 4 | 940 | XP_016861961.1 | |
| XM_017006473.1 | 4 | 889 | XP_016861962.1 | |
| AF172084.1 | 1 | 194 | AAQ13607.1 | |
| AK025953.1 | 1 | 101 | ||
| AK127437.1 | 1 | 1852 | ||
| BC064420.1 | 3 | 1004 | ||
| BC107783.1 | 3 | 988 | AAI07784.1 | |
| BC113456.1 | 3 | 809 | ||
| BC113458.1 | 3 | 806 | ||
| BM723621.1 | 1 | 291 | ||
| CR749364.1 | 3 | 5790 | CAH18217.1 |
Target Gene Details
Entrez Gene ID: | 100506826 |
Gene Name: | MYLK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:123585513-123630821 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK-AS1 | NR_038266.2 | |||
| NR_121654.1 | ||||
| DA531919.1 | ||||
| DA714112.1 | ||||
| DB499540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460837 | Chr.3:123608828 - 124139902 on Build GRCh38 | Gain |
|
| nsv829708 | Chr.3:123519589 - 123690957 on Build GRCh38 | Gain |
|
| nsv829709 | Chr.3:123554281 - 123748120 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map