Assay Details
Target Gene Details
Entrez Gene ID: | 1207 |
Gene Name: | chloride nucleotide-sensitive channel 1A |
Gene Aliases: |
CLCI, CLNS1B, ICln |
Location: |
Chr.11:77616151-77637806 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLNS1A | NM_001293.2 | 7 | 1208 | NP_001284.1 |
| NM_001311200.1 | 6 | 1106 | NP_001298129.1 | |
| NM_001311201.1 | 6 | 1034 | NP_001298130.1 | |
| NM_001311202.1 | 5 | 998 | NP_001298131.1 | |
| AA757599.1 | 1 | 171 | ||
| AF005422.1 | 7 | 1146 | AAB61444.1 | |
| BC119634.1 | ||||
| BC119635.1 | ||||
| BU538016.1 | ||||
| U17899.1 | 7 | 1190 | AAC50111.1 | |
| U53454.1 | 7 | 1200 | AAB03316.1 | |
| X91788.1 | 7 | 1204 | CAA62902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1234n100 | Chr.11:77572991 - 77724786 on Build GRCh38 | Gain |
|
| nsv555452 | Chr.11:77590296 - 77709269 on Build GRCh38 | Gain |
|
| esv3626888 | Chr.11:77610862 - 77622595 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79950774] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map