Assay Details
Target Gene Details
Entrez Gene ID: | 60529 |
Gene Name: | ALX homeobox 4 |
Gene Aliases: |
CRS5, FND2 |
Location: |
Chr.11:44260728-44310166 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALX4 | NM_021926.3 | NP_068745.2 | ||
| AB058691.1 | ||||
| AF294629.1 | AAG23961.1 | |||
| AJ404888.1 | CAC15060.1 | |||
| AK092346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832139 | Chr.11:44063681 - 44277647 on Build GRCh38 | Loss |
|
| dgv67n21 | Chr.11:44250972 - 44277071 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map