Assay Details
Target Gene Details
Entrez Gene ID: | 11080 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B4 |
Gene Aliases: |
DNAJW, DjB4, HLJ1 |
Location: |
Chr.1:77979158-78017964 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB4 | NM_001317100.1 | NP_001304029.1 | ||
| NM_001317101.1 | NP_001304030.1 | |||
| NM_001317102.1 | NP_001304031.1 | |||
| DC412631.1 |
Target Gene Details
Entrez Gene ID: | 8880 |
Gene Name: | far upstream element binding protein 1 |
Gene Aliases: |
FBP, FUBP, hDH V |
Location: |
Chr.1:77944055-77979494 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FUBP1 | NM_001303433.1 | 1 | 100 | NP_001290362.1 |
| NM_003902.4 | 1 | 100 | NP_003893.2 | |
| NR_130152.1 | 1 | 100 | ||
| XM_011542392.2 | XP_011540694.1 | |||
| XM_011542393.2 | XP_011540695.1 | |||
| XM_017002739.1 | XP_016858228.1 | |||
| XM_017002741.1 | XP_016858230.1 | |||
| XM_017002742.1 | XP_016858231.1 | |||
| XM_017002743.1 | XP_016858232.1 | |||
| XM_017002744.1 | XP_016858233.1 | |||
| BQ649331.1 | ||||
| DA113820.1 | 1 | 100 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518855 | Chr.1:77950984 - 77990671 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140681455] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map