Assay Details
Target Gene Details
Entrez Gene ID: | 91544 |
Gene Name: | UBX domain protein 11 |
Gene Aliases: |
COA-1, PP2243, SOC, SOCI, UBXD5 |
Location: |
Chr.1:26282282-26318265 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN11 | NM_001077262.1 | 10 | 1039 | NP_001070730.1 |
| NM_145345.2 | 14 | 1519 | NP_663320.2 | |
| NM_183008.2 | 15 | 1524 | NP_892120.2 | |
| AF370425.1 | 4 | 1312 | AAQ15261.1 | |
| AF520989.1 | 13 | 1228 | AAM76060.1 | |
| AF521017.1 | 13 | 1229 | AAM74201.1 | |
| AF521018.1 | 13 | 1216 | AAM74202.1 | |
| AK074358.1 | 9 | 2033 | ||
| AK090792.1 | 9 | 1998 | ||
| AK093258.1 | 15 | 1468 | BAC04112.1 | |
| AK094198.1 | 10 | 971 | BAC04307.1 | |
| AK097368.1 | 14 | 1617 | BAC05024.1 | |
| AY007163.1 | 11 | 983 | ||
| BX647774.1 | 13 | 2313 | ||
| BX648412.1 | 12 | 2214 | CAH56155.1 | |
| CR749413.1 | 13 | 2347 | CAH18255.1 |
More Information
Set Membership: |
|

Genomic Map