Assay Details
Target Gene Details
Entrez Gene ID: | 387700 |
Gene Name: | solute carrier family 16 member 12 |
Gene Aliases: |
CJMG, CRT2, MCT12 |
Location: |
Chr.10:89430294-89536028 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A12 | NM_213606.3 | 8 | 2306 | NP_998771.3 |
| XM_017016237.1 | 7 | 2396 | XP_016871726.1 | |
| XM_017016238.1 | 7 | 2335 | XP_016871727.1 | |
| XM_017016239.1 | 7 | 2194 | XP_016871728.1 | |
| AK124901.1 | 4 | 1585 | BAC85987.1 | |
| AK127303.1 | 8 | 2307 | BAC86925.1 | |
| BC086873.1 | 3 | 1259 | AAH86873.1 | |
| CR936657.1 | 6 | 4504 |
More Information
Set Membership: |
|

Genomic Map