Assay Details
Target Gene Details
Entrez Gene ID: | 51081 |
Gene Name: | mitochondrial ribosomal protein S7 |
Gene Aliases: |
MRP-S, MRP-S7, RP-S7, RPMS7, S7mt, bMRP27a |
Location: |
Chr.17:75261668-75266376 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS7 | NM_015971.3 | 4 | 634 | NP_057055.2 |
| AF077042.1 | 4 | 583 | AAD27775.1 | |
| AK222995.1 | 4 | 598 | BAD96715.1 | |
| AK313854.1 | 4 | 408 | ||
| BC000241.1 | 4 | 593 | AAH00241.1 | |
| BU599633.1 | 4 | 612 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map