Assay Details
Target Gene Details
Entrez Gene ID: | 30008 |
Gene Name: | EGF containing fibulin like extracellular matrix protein 2 |
Gene Aliases: |
ARCL1B, FBLN4, MBP1, UPH1 |
Location: |
Chr.11:65866441-65872934 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFEMP2 | NM_016938.4 | 7 | 942 | NP_058634.4 |
| NR_037718.1 | 7 | 942 | ||
| AB030655.1 | 7 | 779 | BAA92880.1 | |
| AB209121.1 | 4 | 3018 | BAD92358.1 | |
| AF093119.1 | 6 | 671 | AAC62108.1 | |
| AF109121.1 | 7 | 770 | AAF65188.1 | |
| AF124486.1 | 6 | 683 | AAG45245.1 | |
| AJ132819.2 | 6 | 683 | CAA10791.2 | |
| AK000980.1 | 7 | 779 | ||
| AK075453.1 | 7 | 748 | ||
| AK095794.1 | ||||
| AK098186.1 | 3 | 1682 | ||
| AK292079.1 | 7 | 787 | ||
| AK315181.1 | 7 | 748 | ||
| AY358899.1 | 7 | 744 | AAQ89258.1 | |
| BC010456.1 | 7 | 798 | AAH10456.1 | |
| BQ478413.1 | 4 | 362 | ||
| CR541934.1 | 6 | 683 | CAG46732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| esv999644 | Chr.11:65868696 - 65878563 on Build GRCh38 | Deletion |
|
| nsv555215 | Chr.11:65814108 - 65909045 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map