Assay Details
Target Gene Details
Entrez Gene ID: | 55290 |
Gene Name: | BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit |
Gene Aliases: |
BRFU, TFIIIB50 |
Location: |
Chr.8:37843880-37849913 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRF2 | NM_018310.3 | 3 | 478 | NP_060780.2 |
| AF206673.2 | 3 | 478 | AAG35669.2 | |
| AF298153.1 | 3 | 401 | AAG30222.1 | |
| AK001914.1 | 3 | 440 | BAA91975.1 | |
| AK294337.1 | 4 | 437 | ||
| AK315420.1 | 3 | 427 | ||
| BC010648.1 | 3 | 431 | AAH10648.1 | |
| DA688249.1 | 3 | 478 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509261 | Chr.8:37767151 - 37881434 on Build GRCh38 | Insertion |
|
| nsv528598 | Chr.8:37803553 - 37859603 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map