Assay Details
Target Gene Details
Entrez Gene ID: | 79368 |
Gene Name: | Fc receptor like 2 |
Gene Aliases: |
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C |
Location: |
Chr.1:157745733-157777472 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL2 | NM_001159488.1 | NP_001152960.1 | ||
| NM_030764.3 | NP_110391.2 | |||
| NR_125358.1 | ||||
| XM_011509974.2 | XP_011508276.1 | |||
| XM_011509975.2 | XP_011508277.1 | |||
| XM_011509976.2 | XP_011508278.1 | |||
| XM_017002316.1 | XP_016857805.1 | |||
| XM_017002317.1 | XP_016857806.1 | |||
| XM_017002318.1 | XP_016857807.1 | |||
| XM_017002319.1 | XP_016857808.1 | |||
| AF319438.1 | AAK01402.1 | |||
| AF319439.1 | AAK01403.1 | |||
| AF319440.1 | 1 | 684 | AAK01404.1 | |
| AF390037.1 | AAM12152.1 | |||
| AF459633.1 | AAL60249.1 | |||
| AK301113.1 | ||||
| AL833194.1 | ||||
| AY043465.1 | AAK91778.1 | |||
| AY358130.1 | AAQ88497.1 | |||
| BC069185.1 | AAH69185.1 | |||
| BC129836.1 |
More Information
Set Membership: |
|

Genomic Map