Assay Details
Target Gene Details
Entrez Gene ID: | 22 |
Gene Name: | ATP binding cassette subfamily B member 7 |
Gene Aliases: |
ABC7, ASAT, Atm1p, EST140535 |
Location: |
Chr.X:75053172-75156340 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB7 | NM_001271696.1 | 6 | 807 | NP_001258625.1 |
| NM_001271697.1 | 5 | 687 | NP_001258626.1 | |
| NM_001271698.1 | 5 | 729 | NP_001258627.1 | |
| NM_001271699.1 | 5 | 690 | NP_001258628.1 | |
| NM_004299.4 | 6 | 810 | NP_004290.2 | |
| AF038950.1 | 6 | 747 | AAC39865.1 | |
| AF078777.1 | 6 | 727 | AAD47141.1 | |
| AF133659.1 | 6 | 739 | AAD33045.1 | |
| AK001418.1 | 6 | 767 | ||
| AK294657.1 | 5 | 671 | ||
| AK307414.1 | 5 | 610 | ||
| BC006323.2 | 6 | 743 | AAH06323.1 | |
| BT009918.1 | 6 | 742 | AAP88920.1 | |
| BX537833.1 | 5 | 626 | CAD97847.1 | |
| KU177872.1 | 6 | 742 | ||
| KU177873.1 | 6 | 742 | ||
| KU177874.1 | ||||
| KU177875.1 |
More Information
Set Membership: |
|

Genomic Map