Assay Details
Target Gene Details
Entrez Gene ID: | 113402 |
Gene Name: | SFT2 domain containing 1 |
Gene Aliases: |
C6orf83, pRGR1 |
Location: |
Chr.6:166319728-166342606 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFT2D1 | NM_145169.2 | 8 | 707 | NP_660152.1 |
| NR_130112.1 | 9 | 730 | ||
| NR_130113.1 | 7 | 635 | ||
| NR_130114.1 | 7 | 626 | ||
| AF041429.1 | 8 | 641 | AAF18564.1 | |
| AK021489.1 | 2 | 1974 | ||
| AK090795.1 | 1 | 1615 | ||
| AL833217.1 | 2 | 3038 | ||
| AW964442.1 | 7 | 557 | ||
| BC018969.1 | 8 | 612 | AAH18969.1 | |
| BC028388.1 | 8 | 2325 | AAH28388.1 | |
| BF223532.1 | ||||
| BI597156.1 |
More Information
Set Membership: |
|

Genomic Map