Assay Details
Target Gene Details
Entrez Gene ID: | 666 |
Gene Name: | BOK, BCL2 family apoptosis regulator |
Gene Aliases: |
BCL2L9, BOKL |
Location: |
Chr.2:241558731-241574138 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BOK | NM_032515.4 | 5 | 996 | NP_115904.1 |
| XM_011511696.2 | 5 | 800 | XP_011509998.1 | |
| XM_017004775.1 | 6 | 1098 | XP_016860264.1 | |
| XM_017004776.1 | 6 | 1024 | XP_016860265.1 | |
| AF174487.1 | 5 | 845 | AAD51719.1 | |
| BX372066.2 | 2 | 279 | ||
| BX460019.2 | ||||
| CN279175.1 | 2 | 289 | ||
| H17007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834588 | Chr.2:241498269 - 241613859 on Build GRCh38 | Loss |
|
| nsv584970 | Chr.2:241569597 - 241603273 on Build GRCh38 | Loss |
|
| nsv955299 | Chr.2:241565686 - 241573285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map