Assay Details
Target Gene Details
Entrez Gene ID: | 132 |
Gene Name: | adenosine kinase |
Gene Aliases: |
AK |
Location: |
Chr.10:74151185-74709303 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADK | NM_001202450.1 | 1 | 115 | NP_001189379.1 |
| NM_006721.3 | 1 | 115 | NP_006712.2 | |
| XM_017015698.1 | 1 | 115 | XP_016871187.1 | |
| XM_017015700.1 | 1 | 115 | XP_016871189.1 | |
| XM_017015704.1 | 1 | 115 | XP_016871193.1 | |
| XM_017015705.1 | 1 | 115 | XP_016871194.1 | |
| AK301590.1 | 1 | 98 | ||
| CB133126.1 | 1 | 93 | ||
| DA356691.1 | 1 | 115 | ||
| U33936.1 | AAB01689.1 | |||
| U90338.1 | 1 | 71 | AAB50234.1 |
More Information
Set Membership: |
|

Genomic Map