Assay Details
Target Gene Details
Entrez Gene ID: | 9276 |
Gene Name: | coatomer protein complex subunit beta 2 |
Gene Aliases: |
beta'-COP |
Location: |
Chr.3:139355595-139389680 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPB2 | NM_004766.2 | 16 | 2052 | NP_004757.1 |
| NR_023350.1 | 16 | 2131 | ||
| XM_017007512.1 | 17 | 2237 | XP_016863001.1 | |
| XM_017007513.1 | 15 | 1798 | XP_016863002.1 | |
| AK302943.1 | 16 | 1965 | ||
| BC000326.2 | 16 | 1998 | AAH00326.1 | |
| X70476.1 | 16 | 1990 | CAA49900.1 |
More Information
Set Membership: |
|

Genomic Map