Assay Details
Target Gene Details
Entrez Gene ID: | 5780 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 9 |
Gene Aliases: |
MEG2, PTPMEG2 |
Location: |
Chr.15:75467121-75579291 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN9 | NM_002833.3 | NP_002824.1 | ||
| BC010863.1 | AAH10863.1 | |||
| BC071574.1 | AAH71574.1 | |||
| BT007405.1 | AAP36073.1 | |||
| M83738.1 | AAA60226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map