Assay Details
Target Gene Details
Entrez Gene ID: | 10495 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 2 |
Gene Aliases: |
APK1, COVA1, tNOX |
Location: |
Chr.X:130622330-130903317 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENOX2 | NM_001281736.1 | 14 | 3706 | NP_001268665.1 |
| NM_006375.3 | 15 | 3850 | NP_006366.2 | |
| NM_182314.2 | 16 | 3939 | NP_872114.1 | |
| XM_011531244.2 | 16 | 3941 | XP_011529546.1 | |
| XM_011531245.2 | 16 | 3935 | XP_011529547.1 | |
| XM_017029229.1 | 15 | 3844 | XP_016884718.1 | |
| AF207881.2 | 13 | 3460 | AAF20934.2 | |
| AI540639.1 | ||||
| BC140874.1 | 14 | 3620 |
More Information
Set Membership: |
|

Genomic Map