Assay Details
Target Gene Details
Entrez Gene ID: | 387509 |
Gene Name: | G protein-coupled receptor 153 |
Gene Aliases: |
PGR1 |
Location: |
Chr.1:6247346-6260975 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR153 | NM_207370.2 | 2 | 261 | NP_997253.2 |
| XM_011541434.2 | 2 | 365 | XP_011539736.1 | |
| XM_017001250.1 | 1 | 347 | XP_016856739.1 | |
| XM_017001251.1 | 2 | 250 | XP_016856740.1 | |
| BC068275.1 | 2 | 261 | AAH68275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8n67 | Chr.1:6220621 - 6264303 on Build GRCh38 | Gain |
|
| nsv965 | Chr.1:6253120 - 6297822 on Build GRCh38 | Deletion |
|
| nsv526371 | Chr.1:6247195 - 6307159 on Build GRCh38 | Loss |
|
| nsv831425 | Chr.1:6129291 - 6264418 on Build GRCh38 | Loss |
|
| nsv545236 | Chr.1:6161915 - 6262876 on Build GRCh38 | Loss |
|
| nsv545238 | Chr.1:6248510 - 6257923 on Build GRCh38 | Loss |
|
| nsv508825 | Chr.1:6221275 - 6286549 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map