Assay Details
Target Gene Details
Entrez Gene ID: | 149708 |
Gene Name: | WAP four-disulfide core domain 5 |
Gene Aliases: |
PRG5, WAP1, dJ211D12.5 |
Location: |
Chr.20:45109452-45116321 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC5 | NM_145652.3 | NP_663627.1 | ||
| XM_011528601.1 | XP_011526903.1 | |||
| XM_011528602.1 | XP_011526904.1 | |||
| AY038181.1 | AAK72468.1 | |||
| AY358822.1 | AAQ89181.1 | |||
| BC039173.1 | AAH39173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map