Assay Details
Target Gene Details
Entrez Gene ID: | 55238 |
Gene Name: | solute carrier family 38 member 7 |
Gene Aliases: |
SNAT7 |
Location: |
Chr.16:58665109-58684777 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A7 | NM_001308384.1 | NP_001295313.1 | ||
| NM_018231.2 | 9 | 1380 | NP_060701.1 | |
| XM_017023393.1 | 8 | 1861 | XP_016878882.1 | |
| XM_017023394.1 | XP_016878883.1 | |||
| XM_017023395.1 | 9 | 1356 | XP_016878884.1 | |
| XM_017023396.1 | XP_016878885.1 | |||
| XM_017023397.1 | 9 | 1356 | XP_016878886.1 | |
| AK001677.1 | 9 | 1373 | BAA91830.1 | |
| AK022786.1 | 9 | 1346 | BAB14244.1 | |
| AK222932.1 | 9 | 1342 | BAD96652.1 | |
| AK299424.1 | 9 | 1005 | ||
| BC001961.1 | 8 | 1329 | AAH01961.1 | |
| CR457236.1 | 7 | 980 | CAG33517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509623 | Chr.16:58612215 - 58731990 on Build GRCh38 | Insertion |
|
| esv2758650 | Chr.16:58545101 - 58842133 on Build GRCh38 | Loss |
|
| nsv528513 | Chr.16:58638859 - 58683970 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map