Assay Details
Target Gene Details
Entrez Gene ID: | 7994 |
Gene Name: | lysine acetyltransferase 6A |
Gene Aliases: |
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220 |
Location: |
Chr.8:41929479-42052026 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT6A | NM_006766.4 | 17 | 7533 | NP_006757.2 |
| XM_011544656.2 | 19 | 7759 | XP_011542958.1 | |
| XM_011544657.2 | 18 | 7626 | XP_011542959.1 | |
| XM_011544658.2 | 19 | 7715 | XP_011542960.1 | |
| XM_011544659.2 | 19 | 7738 | XP_011542961.1 | |
| XM_017013863.1 | 18 | 7627 | XP_016869352.1 | |
| XM_017013864.1 | 18 | 7583 | XP_016869353.1 | |
| U47742.1 | 17 | 7475 | AAC50662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831300 | Chr.8:41818817 - 41998976 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map