Assay Details
Target Gene Details
Entrez Gene ID: | 80705 |
Gene Name: | testis specific 10 |
Gene Aliases: |
CEP4L, CT79 |
Location: |
Chr.2:98997261-99154962 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834310 | Chr.2:98917369 - 99093536 on Build GRCh38 | Gain+Loss |
|
| nsv2844 | Chr.2:98977599 - 99022298 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map