Assay Details
Target Gene Details
Entrez Gene ID: | 54552 |
Gene Name: | G protein nucleolar 3 like |
Gene Aliases: |
GNL3B |
Location: |
Chr.X:54530211-54567287 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNL3L | NM_001184819.1 | NP_001171748.1 | ||
| NM_019067.5 | NP_061940.1 | |||
| AK001475.1 | BAA91712.1 | |||
| BC003603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map