Assay Details
Target Gene Details
Entrez Gene ID: | 154043 |
Gene Name: | CNKSR family member 3 |
Gene Aliases: |
MAGI1 |
Location: |
Chr.6:154405299-154510619 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNKSR3 | NM_173515.2 | 13 | 2374 | NP_775786.2 |
| XM_011535485.2 | 13 | 4857 | XP_011533787.1 | |
| AK055911.1 | 13 | 2414 | BAB71044.1 | |
| AK309680.1 | 13 | 1980 | ||
| AY328891.1 | 13 | 2374 | AAQ92306.1 | |
| AY328892.1 | 13 | 2374 | AAQ92307.1 | |
| AY328893.1 | 13 | 2374 | AAQ92308.1 | |
| AY328894.1 | 10 | 1657 | AAQ92309.1 | |
| BC060761.1 | 13 | 2296 | AAH60761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759480 | Chr.6:154389602 - 154436742 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map