Assay Details
Target Gene Details
Entrez Gene ID: | 254187 |
Gene Name: | testis specific 10 interacting protein |
Gene Aliases: |
FAM161C |
Location: |
Chr.11:65945445-65960437 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSGA10IP | NM_152762.2 | 3 | 795 | NP_689975.2 |
| XM_011544887.2 | 3 | 994 | XP_011543189.1 | |
| XM_011544888.2 | 3 | 792 | XP_011543190.1 | |
| AK057442.1 | 3 | 795 | BAB71488.1 | |
| BC104022.1 | 3 | 795 | AAI04023.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map