Assay Details
Target Gene Details
Entrez Gene ID: | 6614 |
Gene Name: | sialic acid binding Ig like lectin 1 |
Gene Aliases: |
CD169, SIGLEC-1, SN, dJ1009E24.1 |
Location: |
Chr.20:3686970-3712558 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIGLEC1 | NM_023068.3 | 21 | 6631 | NP_075556.1 |
| XM_006723610.3 | 22 | 6826 | XP_006723673.1 | |
| XM_011529324.2 | 22 | 6823 | XP_011527626.1 | |
| XM_011529325.2 | 22 | 6829 | XP_011527627.1 | |
| XM_011529326.2 | 22 | 6825 | XP_011527628.1 | |
| XM_011529327.2 | 21 | 6759 | XP_011527629.1 | |
| XM_011529328.2 | 22 | 6771 | XP_011527630.1 | |
| XM_011529329.1 | 20 | 7939 | XP_011527631.1 | |
| XM_011529330.1 | 20 | 7754 | XP_011527632.1 | |
| XM_011529331.1 | 21 | 7930 | XP_011527633.1 | |
| XM_011529332.1 | 20 | 6216 | XP_011527634.1 | |
| AK024459.1 | 5 | 4237 | BAB15749.1 | |
| AK024462.1 | 13 | 4435 | BAB15752.1 | |
| AK024479.1 | 5 | 4237 | BAB15769.1 | |
| AK057560.1 | 3 | 1944 | BAB71527.1 | |
| AK122586.1 | 7 | 7768 | BAC56927.1 | |
| AL833688.1 | 6 | 2366 | ||
| CD365566.1 | 1 | 106 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575355 | Chr.20:3667953 - 3752840 on Build GRCh38 | Gain |
|
| nsv520565 | Chr.20:3652593 - 3740054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map