Assay Details
Target Gene Details
Entrez Gene ID: | 148327 |
Gene Name: | cAMP responsive element binding protein 3 like 4 |
Gene Aliases: |
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL |
Location: |
Chr.1:153967679-153974364 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L4 | NM_001255978.1 | 3 | 582 | NP_001242907.1 |
| NM_001255979.1 | 3 | 368 | NP_001242908.1 | |
| NM_001255980.1 | 3 | 522 | NP_001242909.1 | |
| NM_001255981.1 | 3 | 308 | NP_001242910.1 | |
| NM_130898.3 | 3 | 602 | NP_570968.1 | |
| NR_045658.1 | ||||
| XM_006711172.2 | 3 | 490 | XP_006711235.1 | |
| XM_017000372.1 | 3 | 280 | XP_016855861.1 | |
| AB052778.1 | 3 | 520 | BAC45035.1 | |
| AF394167.1 | 3 | 330 | AAL76113.1 | |
| AF468007.1 | 3 | 369 | AAO33070.1 | |
| AK297703.1 | ||||
| AK304259.1 | 3 | 314 | ||
| AK304665.1 | 3 | 447 | ||
| AK309767.1 | 2 | 743 | ||
| AY049977.1 | 3 | 518 | AAL13157.1 | |
| BC038962.1 | 3 | 504 | AAH38962.1 | |
| BX647716.1 | 3 | 267 |
More Information
Set Membership: |
|

Genomic Map