Assay Details
Target Gene Details
Entrez Gene ID: | 9246 |
Gene Name: | ubiquitin conjugating enzyme E2 L6 |
Gene Aliases: |
RIG-B, UBCH8 |
Location: |
Chr.11:57551655-57568330 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2L6 | NM_004223.4 | NP_004214.1 | ||
| NM_198183.2 | NP_937826.1 | |||
| AF031141.1 | AAB86433.1 | |||
| AF061736.1 | AAD17525.1 | |||
| AK093462.1 | ||||
| AK129621.1 | ||||
| AK226165.1 | ||||
| AK315755.1 | ||||
| BC032491.1 | AAH32491.1 | |||
| CR457126.1 | CAG33407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map