Assay Details
Target Gene Details
Entrez Gene ID: | 54058 |
Gene Name: | chromosome 21 open reading frame 58 |
Gene Aliases: |
- |
Location: |
Chr.21:46300426-46323899 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C21orf58 | NM_001286462.1 | 5 | 786 | NP_001273391.1 |
| NM_001286463.1 | 4 | 597 | NP_001273392.1 | |
| NM_001286476.1 | 5 | 1859 | NP_001273405.1 | |
| NM_001286477.1 | 5 | 1238 | NP_001273406.1 | |
| NM_058180.4 | NP_478060.2 | |||
| XM_005261148.4 | XP_005261205.1 | |||
| XM_006724018.3 | XP_006724081.1 | |||
| XM_011529619.2 | XP_011527921.1 | |||
| XM_011529620.2 | XP_011527922.1 | |||
| XM_011529621.2 | XP_011527923.1 | |||
| XM_011529623.2 | XP_011527925.1 | |||
| XM_011529624.2 | XP_011527926.1 | |||
| XM_011529625.2 | XP_011527927.1 | |||
| XM_011529626.2 | 6 | 3944 | XP_011527928.1 | |
| XM_011529627.2 | 6 | 4024 | XP_011527929.1 | |
| XM_017028388.1 | XP_016883877.1 | |||
| XM_017028389.1 | 6 | 3944 | XP_016883878.1 | |
| XM_017028390.1 | 6 | 1942 | XP_016883879.1 | |
| XM_017028391.1 | 5 | 1765 | XP_016883880.1 | |
| XM_017028392.1 | 7 | 962 | XP_016883881.1 | |
| XM_017028393.1 | 6 | 761 | XP_016883882.1 | |
| AK056387.1 | ||||
| AK096641.1 | 4 | 1514 | ||
| AK098098.1 | 4 | 534 | BAC05232.1 | |
| AY039243.1 | AAK72408.1 | |||
| AY039244.1 | AAK72409.1 | |||
| BC121007.1 | ||||
| BC121008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv3547 | Chr.21:46283787 - 46321516 on Build GRCh38 | Insertion |
|
| nsv527355 | Chr.21:46258860 - 46443112 on Build GRCh38 | Gain |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv587975 | Chr.21:46241321 - 46410929 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|

Genomic Map