Assay Details
Target Gene Details
Entrez Gene ID: | 8878 |
Gene Name: | sequestosome 1 |
Gene Aliases: |
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B |
Location: |
Chr.5:179806388-179838078 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SQSTM1 | NM_001142298.1 | NP_001135770.1 | ||
| NM_001142299.1 | NP_001135771.1 | |||
| NM_003900.4 | NP_003891.1 | |||
| XM_017010010.1 | XP_016865499.1 | |||
| AK025146.1 | ||||
| AK098077.1 | 1 | 510 | ||
| AK226167.1 | ||||
| AK293509.1 | ||||
| AK304877.1 | ||||
| AK312451.1 | ||||
| BC000951.2 | AAH00951.1 | |||
| BC001874.1 | AAH01874.1 | |||
| BC003139.1 | AAH03139.1 | |||
| BC017222.1 | AAH17222.1 | |||
| BC019111.1 | AAH19111.1 | |||
| BI549805.1 | ||||
| DA541885.1 | ||||
| DC398116.1 | ||||
| U41806.1 | AAA93299.1 | |||
| U46751.1 | AAC52070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
| nsv823361 | Chr.5:179786323 - 179824243 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map