Assay Details
Target Gene Details
Entrez Gene ID: | 9308 |
Gene Name: | CD83 molecule |
Gene Aliases: |
BL11, HB15 |
Location: |
Chr.6:14117256-14136918 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD83 | NM_001040280.1 | 5 | 1511 | NP_001035370.1 |
| NM_001251901.1 | 5 | 1343 | NP_001238830.1 | |
| NM_004233.3 | 5 | 1514 | NP_004224.1 | |
| AK223315.1 | 5 | 1373 | BAD97035.1 | |
| AK290426.1 | 5 | 1376 | ||
| BC030830.1 | 5 | 1468 | AAH30830.1 | |
| Z11697.1 | 5 | 1346 | CAA77755.1 |
More Information
Set Membership: |
|

Genomic Map