Assay Details
Target Gene Details
Entrez Gene ID: | 22 |
Gene Name: | ATP binding cassette subfamily B member 7 |
Gene Aliases: |
ABC7, ASAT, Atm1p, EST140535 |
Location: |
Chr.X:75053172-75156340 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB7 | NM_001271696.1 | 9 | 1223 | NP_001258625.1 |
| NM_001271697.1 | 8 | 1103 | NP_001258626.1 | |
| NM_001271698.1 | 8 | 1145 | NP_001258627.1 | |
| NM_001271699.1 | 8 | 1106 | NP_001258628.1 | |
| NM_004299.4 | 9 | 1226 | NP_004290.2 | |
| AF038950.1 | 9 | 1163 | AAC39865.1 | |
| AF078777.1 | 9 | 1143 | AAD47141.1 | |
| AF133659.1 | 9 | 1155 | AAD33045.1 | |
| AK001418.1 | 9 | 1183 | ||
| AK294657.1 | 8 | 1087 | ||
| AK307414.1 | 8 | 1026 | ||
| BC006323.2 | 9 | 1159 | AAH06323.1 | |
| BT009918.1 | 9 | 1158 | AAP88920.1 | |
| BX537833.1 | 8 | 1043 | CAD97847.1 | |
| KU177872.1 | 9 | 1158 | ||
| KU177873.1 | 9 | 1158 | ||
| KU177874.1 | ||||
| KU177875.1 |
More Information
Set Membership: |
|

Genomic Map