Assay Details
Target Gene Details
Entrez Gene ID: | 26289 |
Gene Name: | adenylate kinase 5 |
Gene Aliases: |
AK6 |
Location: |
Chr.1:77281908-77559969 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK5 | NM_174858.2 | NP_777283.1 | ||
| XM_005270739.4 | XP_005270796.1 | |||
| XM_017001010.1 | XP_016856499.1 | |||
| XM_017001012.1 | XP_016856501.1 | |||
| AF445193.1 | AAP97322.1 | |||
| AK090967.1 | BAC03558.1 | |||
| BC036666.1 | AAH36666.1 |
More Information
Set Membership: |
|

Genomic Map