Assay Details
Target Gene Details
Entrez Gene ID: | 51130 |
Gene Name: | ankyrin repeat and SOCS box containing 3 |
Gene Aliases: |
ASB-3 |
Location: |
Chr.2:53669980-53787009 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB3 | NM_001201965.1 | 9 | 2333 | NP_001188894.1 |
| NM_016115.4 | 10 | 2167 | NP_057199.1 | |
| NM_145863.2 | 9 | 1958 | NP_665862.1 | |
| BC009569.1 | AAH09569.1 | |||
| DB558848.1 | 1 | 334 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3844n100 | Chr.2:53667392 - 53720644 on Build GRCh38 | Loss |
|
| esv3583829 | Chr.2:53669630 - 53674975 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map