Assay Details
Target Gene Details
Entrez Gene ID: | 23054 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC2, NRC, PRIP, RAP250, TRBP |
Location: |
Chr.20:34714774-34825649 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA6 | NM_001242539.2 | 12 | 3436 | NP_001229468.1 |
| NM_001318240.1 | 16 | 8937 | NP_001305169.1 | |
| NM_014071.4 | 15 | 6707 | NP_054790.2 | |
| XM_011528721.2 | 15 | 6652 | XP_011527023.1 | |
| XM_011528726.2 | 15 | 6619 | XP_011527028.1 | |
| XM_011528727.2 | 15 | 6616 | XP_011527029.1 | |
| XM_017027739.1 | 16 | 6709 | XP_016883228.1 | |
| XM_017027740.1 | 17 | 6828 | XP_016883229.1 | |
| XM_017027741.1 | 16 | 6706 | XP_016883230.1 | |
| XM_017027742.1 | 16 | 6694 | XP_016883231.1 | |
| XM_017027743.1 | 15 | 6560 | XP_016883232.1 | |
| XM_017027744.1 | 15 | 6598 | XP_016883233.1 | |
| XM_017027745.1 | 15 | 6565 | XP_016883234.1 | |
| XM_017027747.1 | 13 | 6086 | XP_016883236.1 | |
| XM_017027750.1 | 15 | 3730 | XP_016883239.1 | |
| XM_017027751.1 | 14 | 3656 | XP_016883240.1 | |
| AF128458.1 | 14 | 6568 | AAF37003.1 | |
| AF171667.1 | 14 | 6510 | AAF71829.1 | |
| AF177388.1 | 14 | 6526 | AAF13595.1 | |
| AF208227.1 | 16 | 8934 | AAF16403.1 | |
| AF245115.1 | 13 | 6425 | AAF78480.1 | |
| AI498955.1 | 1 | 384 | ||
| AK027040.1 | 5 | 2664 | ||
| BC136272.1 | 14 | 6516 | ||
| D80003.2 | 14 | 6503 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066181 | Chr.20:34682373 - 34725674 on Build GRCh38 | Gain |
|
| nsv819225 | Chr.20:34713328 - 34715327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map