Assay Details
Target Gene Details
Entrez Gene ID: | 54658 |
Gene Name: | UDP glucuronosyltransferase family 1 member A1 |
Gene Aliases: |
BILIQTL1, GNT1, HUG-BR1, UDPGT, UDPGT 1-1, UGT1, UGT1A |
Location: |
Chr.2:233760273-233773299 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A1 | NM_000463.2 | 1 | 340 | NP_000454.1 |
| AK290834.1 | 1 | 344 | ||
| AY435136.1 | 1 | 325 | AAR95637.1 | |
| BC128414.1 | 1 | 341 | ||
| BC128415.1 | 1 | 340 | ||
| DQ364247.1 | 1 | 325 | ||
| JQ686667.1 | 1 | 325 | ||
| JQ699637.1 | 1 | 325 | ||
| JQ699638.1 | 1 | 325 | ||
| JQ699639.1 | 1 | 325 | ||
| JQ699640.1 | 1 | 325 | ||
| JQ699641.1 | 1 | 325 | ||
| JQ699642.1 | 1 | 325 | ||
| JQ699643.1 | 1 | 325 | ||
| JQ699644.1 | 1 | 325 | ||
| JQ699645.1 | 1 | 325 | ||
| JQ699646.1 | 1 | 325 | ||
| JQ699647.1 | 1 | 325 | ||
| JQ699649.1 | 1 | 325 | ||
| JQ699650.1 | 1 | 325 | ||
| M57899.1 | 1 | 340 | AAA63195.1 |
Target Gene Details
Entrez Gene ID: | 54575 |
Gene Name: | UDP glucuronosyltransferase family 1 member A10 |
Gene Aliases: |
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J |
Location: |
Chr.2:233636477-233773305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A10 | NM_019075.2 | NP_061948.1 | ||
| AK025403.1 | ||||
| AY435137.1 | AAR95638.1 | |||
| BC020971.1 | AAH20971.1 | |||
| BC053576.1 | AAH53576.1 | |||
| BC069210.1 | AAH69210.2 | |||
| U89508.1 | AAB81537.1 |
Target Gene Details
Entrez Gene ID: | 54659 |
Gene Name: | UDP glucuronosyltransferase family 1 member A3 |
Gene Aliases: |
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C |
Location: |
Chr.2:233729127-233773299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A3 | NM_019093.2 | NP_061966.1 | ||
| AY435138.1 | AAR95639.1 | |||
| DQ364248.1 |
Target Gene Details
Entrez Gene ID: | 54657 |
Gene Name: | UDP glucuronosyltransferase family 1 member A4 |
Gene Aliases: |
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D |
Location: |
Chr.2:233718792-233773299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A4 | NM_007120.2 | NP_009051.1 | ||
| AK313623.1 | ||||
| AY435139.1 | AAR95640.1 | |||
| BC121036.1 | ||||
| BC131623.1 | ||||
| BC139784.1 | ||||
| DQ364249.1 | ||||
| M57951.1 | AAA63196.1 |
Target Gene Details
Entrez Gene ID: | 54579 |
Gene Name: | UDP glucuronosyltransferase family 1 member A5 |
Gene Aliases: |
UDPGT, UDPGT 1-5, UGT1E |
Location: |
Chr.2:233712992-233773299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A5 | NM_019078.1 | NP_061951.1 | ||
| AY435140.1 | AAR95641.1 | |||
| DQ383514.1 |
Target Gene Details
Entrez Gene ID: | 54578 |
Gene Name: | UDP glucuronosyltransferase family 1 member A6 |
Gene Aliases: |
GNT1, HLUGP, HLUGP1, UDPGT, UDPGT 1-6, UGT1, UGT1A6S, UGT1F |
Location: |
Chr.2:233691675-233773305 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A6 | NM_001072.3 | NP_001063.2 | ||
| NM_205862.1 | NP_995584.1 | |||
| AY435141.1 | AAR95642.1 | |||
| BC011409.1 | ||||
| BC019861.1 | AAH19861.1 | |||
| BM688740.1 | ||||
| BM924331.1 | ||||
| DQ364250.1 | ||||
| J04093.1 | AAA61251.1 |
Target Gene Details
Entrez Gene ID: | 54577 |
Gene Name: | UDP glucuronosyltransferase family 1 member A7 |
Gene Aliases: |
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G |
Location: |
Chr.2:233681938-233773299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A7 | NM_019077.2 | NP_061950.2 | ||
| AY435142.1 | AAR95643.1 | |||
| DQ383513.1 | ||||
| U89507.1 | AAB81536.1 |
Target Gene Details
Entrez Gene ID: | 54576 |
Gene Name: | UDP glucuronosyltransferase family 1 member A8 |
Gene Aliases: |
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H |
Location: |
Chr.2:233617645-233773299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A8 | NM_019076.4 | NP_061949.3 | ||
| AF030310.1 | AAB84259.1 | |||
| AF462267.1 | AAL75963.1 | |||
| AF462268.1 | AAL75964.1 | |||
| AK313488.1 | ||||
| AY435143.1 | AAR95644.1 | |||
| DQ364251.1 |
Target Gene Details
Entrez Gene ID: | 54600 |
Gene Name: | UDP glucuronosyltransferase family 1 member A9 |
Gene Aliases: |
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I |
Location: |
Chr.2:233671898-233773305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A9 | NM_021027.2 | NP_066307.1 | ||
| AF056188.1 | AAC31425.1 | |||
| AK313510.1 | ||||
| AY435144.1 | AAR95645.1 | |||
| BC043491.1 | ||||
| BC058844.1 | AAH58844.1 | |||
| DQ364246.1 | ||||
| S55985.1 | AAB19791.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013712 | Chr.2:233649811 - 234587181 on Build GRCh38 | Loss |
|
| nsv3215 | Chr.2:233722477 - 233761630 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map